| Related Categories | Jenco Compound Microscopes, Jenco Microscopes, Labware, Microscopy |
The BC Series Upright Compound microscopes are designed with a durable, cost effective, modern frame with enhanced stability for image documentation. Its ergonomic, single hand focus/stage control increases workflow and minimizes fatigue. Brightfield Microscope Includes: the four objectives in the table, dust cover, immersion oil, two color filters, spare halogen lamp fuse and instruction manual; Phase Microscope includes all the above plus a green filter and phase centering telescope.
• Stainless steel and brass gear train focus mechanism endures high throughput
• Brilliant images resolved by Infinity or 160 mm objectives
• Versatile performance - multiple microscopic methods available, including brightfield, darkfield, phase contrast and simple polarization
Key Specifications
Optical body- Seidentopf design inclined 30°; 55 to 75 mm interpupillary adjustment, 360° rotation
Nosepiece- Quintuple ball bearing, reversed, with positive click stops and smooth operation
Eye pieces- 10X wide field; Focal Length 25 mm, Field Number 20 mm
Mechanical stage- 8.25 in. (209 mm) x 5.5 in. (140 mm) with vernier scale; Low-positioned right hand coaxial control knobs for one hand focusing; removable spring-clip slide holder
Focus mechanism- adjustable tension control to prohibit drift and adjustable up-stop to protect slides, specimen and objective lenses; markings at 0.002mm increments
Dimensions:
Trinocular: (WHD) (in/cm) 7.8 x 16.5 x 10.9 / 20.0 x 41.9 x 27.8
Binocular: (WHD) (in/cm) 7.8 x 16.2 x 10.9 / 20.0 x 40.9 x 27.8
Jenco is a trademark of Jenco International, Inc.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?