Synonym: pipet tips, pipette tips
| Related Categories | ART PCR Pipette Tips, ART self-sealing barrier pipette tips, Aerosol Barrier Pipette Tips, Labware, Liquid Handling Equipment, |
| sterility | non-sterile |
| packaging | pkg of 1000 tips (bulk) |
| measuring range | 5-200 μL volume |
Compatibility
Biohit Proline-Variable 50-200 μl
Capp Aero 200 μl
Eppendorf Research/Reference 20-200 μl
Eppendorf Research Pro 5-100 μl
Eppendorf Response 4850 5-100 μl
Finnpipette 4027/4500 40-200 μl
Mettler-Toledo VoluMate 20-200 μl
Nichiryo BenchMate 20-200 μl
Pipetman P-200 20-200 μl
Rainin EDP 2 20-250 μl
Rainin EDP Plus 20-250 μl
Rainin Pipet Plus 20-200 μl
Rainin Pipetman 20-200 μl
Socorex/Wheaton 822 Series 10-100 μl
Extended length like the Art XLP but draws a full 200 ul
Precision-molded tips with the Art self-sealing barrier. Guaranteed to prevent carryover contamination during aspiration and delivery of samples. Art tips are ideal for use in genetic studies, forensics, general PCR work and radioisotope sampling. Art tips are available in numerous sizes and styles to meet all your liquid handling needs. All Art tips are certified RNase-, DNase-, DNA- and pyrogen-free.
ART is a registered trademark of Molecular BioProducts, Inc.