Z730203 Sigma

Palladium in Heterocyclic Chemistry: A Guide for the Synthetic Chemist, 2nd ed.

the method of choice for the synthesis of a wide range of biaryls and heterobiaryls

DOWNLOAD MSDS (PDF)
  •  ISBN-13 978-0-08-045117-6

Properties

Related Categories Books and Software, Labware, Organic Chemistry Books More...
publication info   J. Li and G. Gribble, Elsevier, 2007, 658 pp., soft cover

Description

General description

Heterocycles are of paramount importance in the pharmaceutical industry and palladium chemistry is one of the most novel and efficient ways of making heterocycles. Today, palladium-catalyzed coupling is the method of choice for the synthesis of a wide range of biaryls and heterobiaryls. The number of applications of palladium chemistry to the syntheses of heterocycles has grown exponentially.

These developments highlight the need for a monograph dedicated solely to the palladium chemistry in heterocycles and this book provides a comprehensive explanation of the subject. The principal aim of the book is to highlight important palladium-mediated reactions of heterocycles with emphasis on the unique characteristics of individual heterocycles. This book comprises a compilation of important preparations of heteroaryl halides, boranes and stannanes for each heterocycle.

Table of Contents

Chapter 1: Introduction
Chapter 2: Pyrroles
Chapter 3: Indoles
Chapter 4: Pyridines
Chapter 5: Thiophenes and Benzo[b]thiophenes
Chapter 6: Furans and Benzo[b]furans
Chapter 7: Thiazoles and Benzothiazoles
Chapter 8: Oxazoles and Benzoxazoles
Chapter 9: Imidazoles
Chapter 10: Pyrazines and Quinoxalines
Chapter 11: Pyrimidines
Chapter 12: Quinolines
Chapter 13: Pyridazines
Chapter 14: Industrial Scale Palladium Chemistry

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?