EMAIL THIS PAGE TO A FRIEND
A0149 Sigma

Antimycin A1

≥90%

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics A-F, Antibiotics by Application, Antifungal,
assay   ≥90%
Mode of action   enzyme | inhibits
antibiotic activity spectrum   fungi
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
NY1503500

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A8674 Antimycin A from Streptomyces sp.

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?