• USA Home
  • A0186 - N-Acylhexosamine Oxidase from Pseudomonas sp.

EMAIL THIS PAGE TO A FRIEND
A0186 Sigma

N-Acylhexosamine Oxidase from Pseudomonas sp.

suspension, ≥20 units/mg protein

Synonym: N-Acyl-D-glucosamine: oxygen 1-oxidoreductase

Purchase

Description

Biochem/physiol Actions

Enyzme converts N-acetyl-D-glucosamine to N-acetyl-D-glucosaminolactone.

Unit Definition

One unit will oxidize 1.0 μmole of N-acetyl-D-glucosamine to N-acetyl-D-glucosaminolactone per minute at pH 6.8 at 37 °C.

Physical form

Suspension in 80% saturated ammonium sulfate.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?