• USA Home
  • A0394 - N6-(6-Aminohexyl)adenosine 3′,5′-diphosphate lithium salt

EMAIL THIS PAGE TO A FRIEND
A0394 Sigma

N6-(6-Aminohexyl)adenosine 3′,5′-diphosphate lithium salt

  • CAS Number 102029-47-0

  • Empirical Formula (Hill Notation) C16H28N6O10P2

  • Molecular Weight 526.38

  •  MDL number MFCD00056981

Purchase

Properties

InChI Key   IFPMERQTLAQAJP-KHXPSBENSA-M
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?