EMAIL THIS PAGE TO A FRIEND
A0429 Sigma

N4-Aminocytidine

≥95%

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   RSSRMDMJEZIUJX-UHFFFAOYSA-N
assay   ≥95%
storage temp.   2-8°C

Description

Biochem/physiol Actions

Potent mutagen of E. coli and Salmonella typhimurium.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
YU8055000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?