• USA Home
  • A0541 - Actin from porcine muscle

EMAIL THIS PAGE TO A FRIEND
A0541 Sigma

Actin from porcine muscle

lyophilized powder

Purchase

Properties

Related Categories Actin, Actin/Actin-binding Proteins, Biochemicals and Reagents, Cell Biology, Cell Signaling and Neuroscience,
form   lyophilized powder
mol wt   mol wt 43 kDa
composition   Protein, ~90% biuret
storage temp.   2-8°C

Description

Physical form

Lyophilized powder containing Tris, ATP and CaCl2

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?