• USA Home
  • A0700 - (1R,3R)-1-Aminocyclopentane-cis-1,3-dicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
A0700 Sigma

(1R,3R)-1-Aminocyclopentane-cis-1,3-dicarboxylic acid

~99% (HPLC), powder

Synonym: (1R,3R)-ACPD

Purchase

Properties

assay   ~99% (HPLC)
form   powder
color   white
solubility   H2O: soluble20 mg/mL

Description

Biochem/physiol Actions

Potent agonist at N-methyl-D-aspartate (NMDA) receptors; induces accumulation of inositol 1,4,5-triphosphate and total inositol phosphates that is inhibited by MK-9-1 and by low extracellular Ca2+. Does not induce secretion of amyloid precursor protein from cerebral cortex slices.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?