• USA Home
  • A0710 - N-Acetyl-Ala-Glu-Tyr-Tyr-Asn-Lys-Gln-Tyr-Leu-Glu-Gln-Thr-Arg-Ala-Glu-Leu-Asp-Thr amide

EMAIL THIS PAGE TO A FRIEND
A0710 Sigma

N-Acetyl-Ala-Glu-Tyr-Tyr-Asn-Lys-Gln-Tyr-Leu-Glu-Gln-Thr-Arg-Ala-Glu-Leu-Asp-Thr amide

≥90% (HPLC)

Synonym: MHC Class II IAs β chain fragment 58-75

  • CAS Number 157505-54-9

  • Empirical Formula (Hill Notation) C100H150N26O35

  • Molecular Weight 2276.41

  •  MDL number MFCD00677319

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥90% (HPLC)
storage temp.   −20°C
Gene Information   rat ... RT1-Bb(309622)

Description

Biochem/physiol Actions

Prevents experimental autoimmune encephalomyelitis in mice

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?