• USA Home
  • A0732 - Anti-ATG9 (C-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A0732 Sigma

Anti-ATG9 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, PBS solution

Synonym: Anti-APG9 autophagy 9-like 1, Anti-APG9L1, Anti-ATG9 autophagy related 9 homolog A, Anti-ATG9A, Anti-Autophagy 9-like 1 protein, Anti-MGD3208

Purchase

Properties

Related Categories AS-ATG, Alphabetical Index, Antibodies, Antibodies for Autophagy, Antibodies for Cell Biology,
species reactivity   mouse (predicted), rat (predicted), human
application(s)   western blot: 2-5 μg/mL using whole extracts of HEK-293T cells expressing human ATG9
clone   polyclonal
concentration   ~1.0 mg/mL
antibody form   affinity isolated antibody
form   PBS solution
mol wt   antigen mol wt ~130 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ATG9A(79065)
mouse ... Atg9a(245860)
rat ... Atg9a(363254)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 811-824 of human ATG9, conjugated to KLH via a cysteine residue. The corresponding sequence differs by one amino acid in mouse and rat.

General description

Atg9 is a transmembrane autophagy protein that moves between TGN (trans Golgi) and Rab9- and Rab7-positive endosomes. Studies have reported that starvation induces the transport of ATG9 to an endosomal peripheral pool. ATG9 is also known to briefly colocalize with Arp2, an actin-related protein that modulates ATG9 dynamics,,. Anti-ATG9 (C-terminal) recognizes human ATG9.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Application

Rabbit anti-ATG9 (C-terminal) antibody has been used for western blotting analysis in Sertoli cells.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?