EMAIL THIS PAGE TO A FRIEND
A0753 Sigma

DL-Ala-Gly

  • CAS Number 1188-01-8

  • Empirical Formula (Hill Notation) C5H10N2O3

  • Molecular Weight 146.14

  •  PubChem Substance ID 329770628

Purchase

Properties

InChI Key   CXISPYVYMQWFLE-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A0878 Ala-Gly

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?