• USA Home
  • A0836 - Acetofluoro-α-D-glucose

EMAIL THIS PAGE TO A FRIEND
A0836 Sigma

Acetofluoro-α-D-glucose

Synonym: 2,3,4,6-Tetra-O-acetyl-α-D-glucopyranosyl fluoride

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates A-C, Monosaccharide More...
InChI Key   JJXATNWYELAACC-RGDJUOJXSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?