• USA Home
  • A1030 - Adenosine Deaminase from calf intestinal mucosa

EMAIL THIS PAGE TO A FRIEND
A1030 Sigma

Adenosine Deaminase from calf intestinal mucosa

Type VIII, ammonium sulfate suspension, ~200 units/mg protein

Synonym: Adenosine aminohydrolase

Purchase

Properties

Related Categories 3.5.x.x Acting on C-N other than peptides, 3.x.x.x Hydrolases, Biochemicals and Reagents, Enzyme Class Index, Enzymes, Inhibitors, and Substrates More...
type   Type VIII
form   ammonium sulfate suspension
storage temp.   2-8°C

Description

Analysis Note

Protein determined by biuret.

Unit Definition

One unit will deaminate 1.0 μmole of adenosine to inosine per min at pH 7.5 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, pH approx. 6

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A5168 Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?