EMAIL THIS PAGE TO A FRIEND
A103 Sigma

L-Aspartic acid

Synonym: (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

  • CAS Number 56-84-8

  • Linear Formula HO2CCH2CH(NH2)CO2H

  • Molecular Weight 133.10

  •  Beilstein Registry Number 1723530

  •  EC Number 200-291-6

  •  MDL number MFCD00002616

  • Popular Documents:  FTIR (PDF)  

Purchase

Properties

InChI Key   CKLJMWTZIZZHCS-REOHCLBHSA-N
mp   >300 °C (dec.)(lit.)

Description

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
CI9098500
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

A9256 L-Aspartic acid, reagent grade, ≥98% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?