• USA Home
  • A1173 - Albumin from porcine serum

EMAIL THIS PAGE TO A FRIEND
A1173 Sigma

Albumin from porcine serum

lyophilized powder, essentially fatty acid free, essentially globulin free, ~99% (agarose gel electrophoresis)

Purchase

Properties

Related Categories Albumin, Albumins from other species, Biochemicals and Reagents, Proteins and Derivatives
assay   ~99% (agarose gel electrophoresis)
form   lyophilized powder
impurities   ≤0.005% Fatty acid
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?