• USA Home
  • A1181 - Ammonia–boranecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
A1181 Sigma

Ammonia–boranecarboxylic acid

Synonym: Carboxyborane ammonia complex

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   MUTDNIUYABKXLN-UHFFFAOYSA-N
storage temp.   −20°C

Description

Biochem/physiol Actions

Hypolipidemic and antineoplastic agent.

Linkage

α-Boron analog of glycine.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
ED1160000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?