• USA Home
  • A1210 - N-Acetyl-Val-Ala-Asp-al

EMAIL THIS PAGE TO A FRIEND
A1210 Sigma

N-Acetyl-Val-Ala-Asp-al

≥98% (TLC)

Purchase

Properties

Related Categories Aldehydes, Biochemicals and Reagents, Building Blocks, C13-C60, Carbonyl Compounds,
InChI Key   BGFYQRSQECIBIB-PEXQALLHSA-N
assay   ≥98% (TLC)
storage temp.   −20°C

Description

Packaging

1, 5 mg in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?