• USA Home
  • A1216 - N-Acetyl-Ile-Glu-Thr-Asp-al

EMAIL THIS PAGE TO A FRIEND
A1216 Sigma

N-Acetyl-Ile-Glu-Thr-Asp-al

~99% (TLC), powder

Synonym: Ac-IETD-CHO

  • Empirical Formula (Hill Notation) C21H34N4O10

  • Molecular Weight 502.52

  •  MDL number MFCD01318858

  •  PubChem Substance ID 329770688

Purchase

Description

Biochem/physiol Actions

Caspase 8 inhibitor; blocks the cleavage of the 32 kDa caspase 3 precursor into the p12 and p20 subunits, thus blocking the formation of active caspase 3.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Price and Availability


Flow Cytometry Instruments

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?