• USA Home
  • A1224 - N-(4-Acetamidophenyl)indomethacinamide

EMAIL THIS PAGE TO A FRIEND
A1224 Sigma

N-(4-Acetamidophenyl)indomethacinamide

≥98%, crystalline

Synonym: N-4AcetIA, N-(4-Acetamidophenyl)-1-(4-chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetamide

  • Empirical Formula (Hill Notation) C27H24N3O4Cl

  • Molecular Weight 489.95

  •  MDL number MFCD02683236

  •  PubChem Substance ID 329770691

Purchase

Properties

Related Categories A to C, Application Index, Arachidonic Acid Cascade, Biochemicals and Reagents, Cancer Research,
InChI Key   CVMUUPNNEPVQCL-UHFFFAOYSA-N
assay   ≥98%
form   crystalline
solubility   DMSO: ≥15 mg/mL
  DMF: ≥23 mg/mL
absorption   λmax 267 nm
shipped in   wet ice
storage temp.   −20°C

Description

Biochem/physiol Actions

Potent and selective reversible COX-2 inhibitor. N-4-AcetIA inhibits human and ovine COX-2 with IC50 of 0.1 μM and 0.625 μM, respectively. About 400 times less potent as an inhibitor of human COX-1 and 80 times less potent as an inhibitor of ovine COX-1. Shows anti−inflammatory, antiangiogenic, and cancer chemopreventive activity in various experimental models.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?