• USA Home
  • A1262 - 8-Azidoadenosine 3′:5′-cyclic monophosphate

EMAIL THIS PAGE TO A FRIEND
A1262 Sigma

8-Azidoadenosine 3′:5′-cyclic monophosphate

~95%

  • CAS Number 31966-52-6

  • Empirical Formula (Hill Notation) C10H11N8O6P

  • Molecular Weight 370.22

  •  MDL number MFCD00057127

  •  PubChem Substance ID 24890518

Purchase

Description

Application

Photoaffinity label for cAMP-dependent enzymes.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?