EMAIL THIS PAGE TO A FRIEND
A1416 Sigma

β-Ala-Phe

  • CAS Number 17136-28-6

  • Empirical Formula (Hill Notation) C12H16N2O3

  • Molecular Weight 236.27

  •  MDL number MFCD00069948

  •  PubChem Substance ID 24890543

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   FLJLAJANCPPHDW-JTQLQIEISA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?