• USA Home
  • A144 - 1-Allyl-3,7-dimethyl-8-phenylxanthine

EMAIL THIS PAGE TO A FRIEND
A144 Sigma

1-Allyl-3,7-dimethyl-8-phenylxanthine

solid

  • CAS Number 149981-23-7

  • Empirical Formula (Hill Notation) C16H16N4O2

  • Molecular Weight 296.32

  •  MDL number MFCD00083166

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   DKISSNPEWQAXRA-UHFFFAOYSA-N
form   solid
color   white
solubility   DMSO: soluble
  H2O: insoluble
  methanol: insoluble

Description

Biochem/physiol Actions

A1 adenosine receptor antagonist.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?