EMAIL THIS PAGE TO A FRIEND
A1503 Sigma

Ala-His

  • CAS Number 3253-17-6

  • Empirical Formula (Hill Notation) C9H14N4O3

  • Molecular Weight 226.23

  •  EC Number 221-845-3

  •  PubChem Substance ID 24890550

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides, Dipeptides and Tripeptides, Peptides
InChI Key   XZWXFWBHYRFLEF-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?