• USA Home
  • A1585 - Amino acid standards, physiological

EMAIL THIS PAGE TO A FRIEND
A1585 Sigma

Amino acid standards, physiological

basics

Purchase

Properties

Related Categories Additional Standards, Amino Acids, Analytical Standards, Analytical/Chromatography, Biochemicals and Reagents,
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   2-8°C

Description

Other Notes

Amino acids and related compounds are 2.5 μmoles per mL except L-anserine and L-homocystine at 1.25 μmole per ml.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Amino Acids Reference Chart

The hydrophobicity index is a measure of the relative hydrophobicity, or how soluble an amino acid is in water. In a protein, hydrophobic amino acids are likely to be found in the interior, whereas h...

Related Products

replaced by

Product #

Description

Add to Cart

A6282 Amino acid standards, physiological, analytical standard, basics

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?