• USA Home
  • A1610 - Anti-AMHR2 (364-375) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
A1610 Sigma

Anti-AMHR2 (364-375) antibody produced in rabbit

IgG fraction of antiserum, PBS solution

Synonym: Anti-AMHR, Anti-Anti-Muellerian hormone type-2 receptor precursor, Anti-MISRII

Purchase

Properties

Related Categories AM-AN, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   PBS solution
mol wt   antigen mol wt ~63 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... AMHR2(269)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 364-375 of human AMHR2

General description

Anti-Mullerian hormone (AMH) signaling is thought to be involved in the onset of natural menopause. Genetic interactions of AMH and AMH receptor 2 (AMHR2) may modify age at natural menopause. Furthermore, AMHR2 gene polymorphisms may be associated with the risk of polycystic ovary syndrome (PCOS). Rabbit anti- AMHR2 (364-375) antibody is specific for AMHR2 (364-375).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Application

Anti-AMHR2 (364-375) antibody can be used for western blot (1:500-1:2,000).

Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about the Sigma Life Science and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?