EMAIL THIS PAGE TO A FRIEND
A1799 Sigma

Acrylamide

Purchase

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2074 6.1 / PGIII
WGK Germany 
3
RTECS 
AS3325000
Flash Point(F) 
280.4 °F
Flash Point(C) 
138 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?