• USA Home
  • A180 - Nω-Nitro-D-arginine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
A180 Sigma

Nω-Nitro-D-arginine methyl ester hydrochloride

Synonym: D-NAME, NG-Nitro-D-arginine methyl ester hydrochloride

  • CAS Number 50912-92-0

  • Empirical Formula (Hill Notation) C7H15N5O4 · HCl

  • Molecular Weight 269.69

  •  MDL number MFCD00133613

  •  PubChem Substance ID 329770723

Purchase

Properties

InChI Key   QBNXAGZYLSRPJK-UHFFFAOYSA-N
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

N4770 Nω-Nitro-D-arginine methyl ester hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?