EMAIL THIS PAGE TO A FRIEND
A1897 Sigma

Aluminum oxide

Type WA-7: Acid

Synonym: Alumina

Purchase

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Aluminum, Aluminum Oxide, Essential Chemicals, Materials Science,
InChI Key   TWNQGVIAIRXVLR-UHFFFAOYSA-N
type   Type WA-7: Acid
mp   2040 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
BD1200000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?