• USA Home
  • A189 - Amyloid β Protein Fragment 1-40

EMAIL THIS PAGE TO A FRIEND
A189 Sigma

Amyloid β Protein Fragment 1-40

Synonym: Aβ40

  • CAS Number 131438-79-4

  • Empirical Formula (Hill Notation) C194H295N53O58S

  • Molecular Weight 4329.80

  •  MDL number MFCD00130509

Purchase

Properties

storage temp.   −20°C

Description

Amino Acid Sequence

Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

A1075 Amyloid β Protein Fragment 1-40, ≥90% (HPLC), powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?