• USA Home
  • A1907 - 5′-Adenylic Acid Deaminase from Aspergillus sp.

EMAIL THIS PAGE TO A FRIEND
A1907 Sigma

5′-Adenylic Acid Deaminase from Aspergillus sp.

lyophilized powder, ~0.1 unit/mg solid

Purchase

Properties

Related Categories Cell Biology, General Metabolic Enzymes, Metabolomics, Nucleotide Metabolism
form   lyophilized powder
composition   Protein, ~20% Lowry
storage temp.   2-8°C

Description

Application

This enzyme is not specific for 5′-AMP. It deaminates adenosine at approx. 50% of the rate with 5′-AMP.

Unit Definition

One unit will deaminate 1.0 μmole of 5′-AMP to 5′-IMP per min at pH 6.5 at 25 °C.

Physical form

Lyophilized powder containing diatomaceous earth

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?