• USA Home
  • A1930 - ACCUSPIN Tubes Nonsterile, 50 mL Capacity
EMAIL THIS PAGE TO A FRIEND
A1930 Sigma
ACCUSPIN Tubes Nonsterile, 50 mL Capacity
Purchase
Properties
Related Categories Centrifugation Media, Core Bioreagents, Research Essentials More...
IVD   for in vitro diagnostic use
Quality Level   GMP-IVD
Description
General description
Polypropylene tube fitted with a high density polyethylene barrier. Each tube will accept 15 mL of density gradient.
Legal Information
Accuspin is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?