EMAIL THIS PAGE TO A FRIEND
A2041 Sigma

D-Ala-Ala-Ala

  • CAS Number 5874-89-5

  • Empirical Formula (Hill Notation) C9H17N3O4

  • Molecular Weight 231.25

  •  MDL number MFCD00069941

  •  PubChem Substance ID 24890620

Purchase

Properties

Related Categories Biochemicals and Reagents, Dipeptides and Tripeptides, Peptides, Tripeptides
InChI Key   BYXHQQCXAJARLQ-UHFFFAOYSA-N
storage temp.   −20°C

Description

Amino Acid Sequence

Ala-Ala-Ala

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?