• USA Home
  • A204 - Anti-Epidermal Growth Factor Receptor antibody produced in sheep

EMAIL THIS PAGE TO A FRIEND
A204 Sigma

Anti-Epidermal Growth Factor Receptor antibody produced in sheep

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-EGFR

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Cytokines and Growth Factors,
species reactivity   rat, dog, human, chicken
application(s)   immunocytochemistry: 1:50-1:200 using formalin-fixed cells
  immunofluorescence: suitable
  immunohistochemistry: suitable
  immunoprecipitation: 10-12 μg/mL using A-431 epidermoid carcinoma cell extracts.
  western blot: 1:50-1:200
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... EGFR(1956)
rat ... Egfr(24329)
biological source   sheep
conjugate   unconjugated

Description

Immunogen

fusion protein (20 amino acid residues) of the human EGF receptor. The sequence is proximal to the phosphorylation region (near the N-terminal sequence).

General description

EGFR is a glycoprotein receptor that consists of 3 domains: an extracellular domain which modulates ligand binding and receptor dimerization, a single transmembrane domain, and a cytoplasmic domain. EGFR/ErbB1 binds to EGF ligands and is known to form homodimers or heterodimers. Furthermore, EGFR regulates differentiation, growth, movement and apoptosis in a wide range of cells.
Sheep anti-EGFR recognizes rat, mouse, dog, chicken and human EGF receptors. Furthermore, the antibody recognizes the internal domain of the receptor molecule and will block the phosphorylation but will not block EGF binding. The antibody can be used with formalin-fixed, paraffin-embedded tissue sections.

Physical form

0.2 μm filtered solution in 0.15 M phosphate buffered saline, pH 7.5.

Preparation Note

Purified by protein G affinity chromatography.

Application

Sheep anti-EGFR antibody was used to detect EGFR by immunofluorescence in mouse oocyte cumulus cells that were previously fixed in 4% paraformaldehyde

Sheep anti-epidermal growth factor receptor (EGFR) can be used for immunohistochemistry, immunoprecipitation, western blot and immunocytochemistry applications.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

E2156 Monoclonal Anti-EGF Receptor antibody produced in mouse, clone 225, purified from hybridoma cell culture
HPA001200 Anti-EGFR antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
E1282 Anti-Epidermal Growth Factor Receptor antibody produced in goat, affinity isolated antibody, lyophilized powder
E1157 Anti-Epidermal Growth Factor Receptor/Erb B1 antibody produced in goat, affinity isolated antibody, lyophilized powder
E0532 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 102618, purified immunoglobulin, lyophilized powder
E2760 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone 29.1, ascites fluid
E3641 Epidermal Growth Factor Receptor human, buffered aqueous glycerol solution, 5,000-30,000 units/mg protein (Lowry)
E3138 Monoclonal Anti-Epidermal Growth Factor Receptor antibody produced in mouse, clone F4, ascites fluid
E2645 Epidermal Growth Factor Receptor human, lyophilized powder, ≥15,000 units/mg protein (Bradford)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?