• USA Home
  • A2135 - Azotobacter vinelandii cells

EMAIL THIS PAGE TO A FRIEND
A2135 Sigma

Azotobacter vinelandii cells

lyophilized cells

Synonym: ATCC 12518

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
sterility   Not processed or packaged aseptically
form   lyophilized cells
storage temp.   −20°C

Description

Other Notes

Not intended for use as a starter culture.

Preparation Note

Produced in pure culture

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?