• USA Home
  • A214 - α2C-Adrenergic Receptor human

EMAIL THIS PAGE TO A FRIEND
A214 Sigma

α2C-Adrenergic Receptor human

membrane suspension, recombinant, expressed in Sf9 cells

Purchase

Properties

recombinant   expressed in Sf9 cells
form   membrane suspension
storage temp.   −70°C
Gene Information   human ... ADRA2C(152)

Description

Analysis Note

Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.

Caution

Membranes retain original activity for several months when stored in original packaging solution.

Packaging

Actual concentrations and specific binding capacity are provided with each lot.

Physical form

Membrane suspension in 50 mM Tris-HCl, pH 7.4, 10% glycerol, 1% BSA

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?