• USA Home
  • A2163 - N-Acetyl-Phe-Nle-Arg-Phe amide

EMAIL THIS PAGE TO A FRIEND
A2163 Sigma

N-Acetyl-Phe-Nle-Arg-Phe amide

≥95% (HPLC)

  • CAS Number 83903-28-0

  • Empirical Formula (Hill Notation) C32H46N8O5

  • Molecular Weight 622.76

  •  MDL number MFCD00076633

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Invertebrate Neuropeptides, Neuropeptides, Peptides and Proteins,
InChI Key   GPNWSJNNYYTLLQ-UHFFFAOYSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

FMRF amide analog; molluscan cardioexcitatory neuropeptide analog.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?