• USA Home
  • A2223 - Axl/Fc Chimera from mouse

EMAIL THIS PAGE TO A FRIEND
A2223 Sigma

Axl/Fc Chimera from mouse

≥90% (SDS-PAGE), recombinant, expressed in NSO cells

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cancer Research, Cell Biology, Cell Signaling Enzymes,
recombinant   expressed in NSO cells
assay   ≥90% (SDS-PAGE)
mol wt   calculated mol wt 72.3 kDa
  mol wt 100-110 kDa by SDS-PAGE (reducing)
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   mouse ... Axl(26362)

Description

Analysis Note

Measured by its ability to bind recombinant human Gas6.

Biochem/physiol Actions

Member of a tyrosine kinase receptor subfamily shown to be involved in hematopoiesis, embryonic development, tumorigenesis, and testicular function regulation.

General description

Extracellular domain of mouse Axl (residues 1-443) fused to the C-terminal histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from a 0.2 μm filtered solution of phosphate buffered saline.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?