• USA Home
  • A2262 - DL-Amphetamine sulfate salt solution

EMAIL THIS PAGE TO A FRIEND
A2262 Sigma

DL-Amphetamine sulfate salt solution

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Additional Standards, Amphetamine, Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography,
grade   drug standard
InChI Key   PYHRZPFZZDCOPH-UHFFFAOYSA-N
assay   ≥98%
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C
Protocols & Articles

Articles

GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN

From our library of Articles, Sigma-Aldrich presents GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX® G-PN
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-DP

From our library of Articles, Sigma-Aldrich presents GC Analysis of Amphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-DP
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

GC Analysis of Amphetamine and Methamphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN

From our library of Articles, Sigma-Aldrich presents GC Analysis of Amphetamine and Methamphetamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN
Keywords: Chromatography, Clinical, Flame ionization detector, Forensic, Gas chromatography, Pharmaceutical, Purification

HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Amphetamine and Methamphetamine Enantiomers on Astec® CHIROBIOTIC® V2
Keywords: Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

A5880 D-Amphetamine hemisulfate salt

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?