• USA Home
  • A2347 - N-Acetyl-Gly(t-butyl)-Gly(t-butyl)-Asn(Methyl)2-Ala-7-amido-4-methylcoumarin

EMAIL THIS PAGE TO A FRIEND
A2347 Sigma

N-Acetyl-Gly(t-butyl)-Gly(t-butyl)-Asn(Methyl)2-Ala-7-amido-4-methylcoumarin

≥97% (HPLC), solid

  • CAS Number 204909-38-6

  • Empirical Formula (Hill Notation) C33H48N6O8

  • Molecular Weight 656.77

  •  MDL number MFCD03452807

Purchase

Description

Amino Acid Sequence

Ac-tBu-Gly-tBu-Gly-Asn(Me)2-Ala-AMC

General description

Short human cytomegalovirus (HCMV) protease substrate.

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?