• USA Home
  • A2422 - D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr amide acetate salt

EMAIL THIS PAGE TO A FRIEND
A2422 Sigma

D-Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr amide acetate salt

≥97% (HPLC)

Synonym: [D-Ala1]-Peptide T amide

Purchase

Properties

Related Categories AIDS and Viral Research, Cell Biology, Cell Signaling and Neuroscience, Immunomodulators, Peptides and Proteins,
InChI Key   RNRQNIXDWUFGFX-ZPGAAPGCSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Biochem/physiol Actions

HIV inhibitor.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?