• USA Home
  • A2424 - Amyloid TISSUE-TROL

EMAIL THIS PAGE TO A FRIEND
A2424 Sigma

Amyloid TISSUE-TROL

from Human heart tissue containing intercellular amyloid

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from Human heart tissue containing intercellular amyloid
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Application

Suitable for use with Sigma Amyloid Procedure No. HT60. 25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained using Sigma Procedure No. HT60.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

TTR002 Amyloid TISSUE-TROL Control Slides, from Human heart tissue containing intercellular amyloid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?