EMAIL THIS PAGE TO A FRIEND
A2460 Sigma

Ammonium dichromate

BioXtra

Synonym: Ammonium bichromate

Purchase

Properties

vapor density   8.7 (vs air)
InChI Key   XAQCJVGGJJFLPP-UHFFFAOYSA-L
product line   BioXtra
impurities   <0.0005% Phosphorus (P)
  <0.1% Insoluble matter
mp   170 °C (dec.)(lit.)
anion traces   chloride (Cl-): <0.005%
  sulfate (SO42-): <0.01%
cation traces   Al: <0.0005%
  Ca: <0.005%
  Cu: <0.0005%
  Fe: <0.001%
  K: <0.01%
  Mg: <0.0005%
  Na: <0.01%
  Pb: <0.0005%
  Zn: <0.0005%

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1439 5.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?