EMAIL THIS PAGE TO A FRIEND
A2461 Sigma

Amberlyst® 131

wet

Purchase

Properties

form   wet
contains   65% moisture content
feature   temperature 250 °C
matrix active group   —SO3- • H+
capacity   1.8 meq/mL exchange capacity
  4.8 meq/g exchange capacity

Description

Application

Strongly acidic Gel ion-exchange catalyst

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?