• USA Home
  • A2464 - β-N-Acetylgalactosaminidase from Bacillus sp.

EMAIL THIS PAGE TO A FRIEND
A2464 Sigma

β-N-Acetylgalactosaminidase from Bacillus sp.

≥10 units/mL, buffered aqueous solution

Synonym: 2-Acetamido-2-deoxy-β-D-galactoside acetamidodeoxygalactohydrolase

Purchase

Properties

form   buffered aqueous solution
concentration   ≥10 units/mL
storage temp.   −20°C

Description

Unit Definition

One unit will release 1.0 μmole of p-nitrophenol from p-nitrophenyl-N-acetyl-β-D-galactosaminide per min at pH 6.0 at 37°C.

Physical form

Solution in 10 mM Tris-HCl, pH 8.0

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?