• USA Home
  • A2465 - Antifungal assay agar

EMAIL THIS PAGE TO A FRIEND
A2465 Sigma

Antifungal assay agar

for microbiology

Purchase

Properties

Related Categories All Solid Media/Agars, Media, Microbiology More...
grade   for microbiology

Description

Components

Ingredients (g/L):
Dextrose, 50.00
Sodium citrate, 4.50
Potassium phosphate, 0.55
Citric acid, 1.00
Casein enzymic hydrolysate, 4.00
Pyridoxine hydrochloride, 0.00025
Thiamine, 0.00025
Inositol, 0.025
Calcium pantothenate, 0.0025
Niacin, 0.0025
Potassium chloride, 0.425
Calcium chloride, 0.125
Magnesium sulfate, 0.125
Ferric chloride, 0.0025
Manganese sulfate, 0.0025
Biotin, 0.000008
Agar, 15.00

Application

Recommended for assaying antifungal activity.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?