EMAIL THIS PAGE TO A FRIEND
A2468 Sigma

Atropine

meets USP testing specifications

Synonym: Hyoscyamine, Tropine tropate, endo-(±)-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester

  • CAS Number 51-55-8

  • Empirical Formula (Hill Notation) C17H23NO3

  • Molecular Weight 289.37

  •  Beilstein Registry Number 91260

  •  EC Number 200-104-8

  •  MDL number MFCD00022622

  •  PubChem Substance ID 24890659

Purchase

Properties

Related Categories Analytical/Chromatography, Pharmacopoeia, Pharmacopoeia A-Z More...
InChI Key   RKUNBYITZUJHSG-SPUOUPEWSA-N
suitability   meets USP testing specifications
Gene Information   human ... CHRM1(1128), CHRM2(1129), CHRM3(1131), CHRM4(1132), CHRM5(1133)
mouse ... Chrm1(12669)
rat ... Chrm1(25229), Chrm2(81645), Chrm3(24260), Drd2(24318), Htr1a(24473)

Description

Biochem/physiol Actions

Competitive nonselective antagonist at central and peripheral muscarinic acetylcholine receptors.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3
RTECS 
CK0700000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?