EMAIL THIS PAGE TO A FRIEND
A2520 Sigma

Acridine Yellow

  •  Colour Index Number 46025

  •  EC Number 205-194-2

Purchase

Properties

composition   Dye content, ~90%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
AR8790000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?