• USA Home
  • A2597 - [pGlu3]-Amyloid β-Protein Fragment 3-42

EMAIL THIS PAGE TO A FRIEND
A2597 Sigma

[pGlu3]-Amyloid β-Protein Fragment 3-42

≥90% (HPLC), solid

  • CAS Number 183449-57-2

  • Empirical Formula (Hill Notation) C196H299N53O55S

  • Molecular Weight 4309.86

  •  MDL number MFCD03457978

Purchase

Description

Amino Acid Sequence

Glp-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala

Biochem/physiol Actions

Predominant amyloid β-peptide structure deposited in human brain of Alzheimer′s disease and Down′s syndrome patients. It is suggested to accumulate in the brain and to trigger the formation of insoluble amyloid β-peptide deposits.

Price and Availability


Amplified Detection

Free Webinar

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?