• USA Home
  • A2635 - N-Acetyl-L-3-nitrotyrosine ethyl ester

EMAIL THIS PAGE TO A FRIEND
A2635 Sigma

N-Acetyl-L-3-nitrotyrosine ethyl ester

  • CAS Number 40642-95-3

  • Empirical Formula (Hill Notation) C13H16N2O6

  • Molecular Weight 296.28

  •  MDL number MFCD00057592

  •  PubChem Substance ID 24890678

  •  eCl@ss 32160406

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   VFDMIQARHVLPBK-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?